Learn More
Kyfora Bio DSPG, Sodium Salt - 500MG
Catalog No. 502559814
Encompass_Preferred
DSPG (1,2-Distearoyl-sn-glycero-3-phospho-rac-glycerol) is an anionic phospholipid widely used in liposomal formulations for drug and gene delivery. The inclusion of DSPG promotes membrane destabilization, which facilitates the release of therapeutic agents from the liposomal core. Liposomes incorporating both anionic and zwitterionic lipids have shown enhanced DNA entrapment efficiency and excellent stability, making DSPG a valuable component in delivery systems for in vitro and in vivo applications. For research use only.Key Applications: Drug delivery, gene delivery
Catalog No. 50-255-9814 Supplier Kyfora Bio Supplier No. 26453500
DSPG (1,2-Distearoyl-sn-glycero-3-phospho-rac-glycerol) is an anionic phospholipid widely used in liposomal formulations for drug and gene delivery. The inclusion of DSPG promotes membrane destabilization, which facilitates the release of therapeutic agents from the liposomal core. Liposomes incorporating both anionic and zwitterionic lipids have shown enhanced DNA entrapment efficiency and excellent stability, making DSPG a valuable component in delivery systems for in vitro and in vivo applications. For research use only.Key Applications: Drug delivery, gene delivery