Learn More
MSC Atrix Dust Sampling Filter, U15 PTFE, Exceeds HEPA, Disposable, 6/Pack
Catalog No. 502815889
Encompass
The ATISF-U15 is an economical, highly efficient, single-use, disposable dust sampling filter manufactured with U15 PTFE (99.9995% Efficiency) Filtration Media. It is independently tested to exceed HEPA (99.97% efficiency at 0.3 Micron) efficiency, designed for immediate containment of particulate, and compatible with various vacuum series for hazardous particulate, pest control, and bulk sampling identification.
  • Sampling filter for identification
  • Immediate containment of particulate
  • Exceeds HEPA (H13) standards
  • ESD safe
  • Cleanroom compatible (Class 100 Cleanroom (ISO 5) or Class 100,000 (ISO 8))
  • Single-use disposable design
  • Clear ABS housing for visual analysis of sampling contents
Catalog No. 50-281-5889 Supplier MSC Supplier No. 16940264
The ATISF-U15 is an economical, highly efficient, single-use, disposable dust sampling filter manufactured with U15 PTFE (99.9995% Efficiency) Filtration Media. It is independently tested to exceed HEPA (99.97% efficiency at 0.3 Micron) efficiency, designed for immediate containment of particulate, and compatible with various vacuum series for hazardous particulate, pest control, and bulk sampling identification.
  • Sampling filter for identification
  • Immediate containment of particulate
  • Exceeds HEPA (H13) standards
  • ESD safe
  • Cleanroom compatible (Class 100 Cleanroom (ISO 5) or Class 100,000 (ISO 8))
  • Single-use disposable design
  • Clear ABS housing for visual analysis of sampling contents