Learn More
Electron Microscopy Sciences EMS Disposable Macro Knives Assorted Pack Standard
SDP
Catalog No. 501931597
Encompass_Preferred
These disposable knives are economically priced and completely disposable. They have double honed, flexible blades with non-slip handles to ensure accurate dissection. They may be stropped for a longer lasting edge. They are available in standard or premium grades of Sheffield stainless steel.
Catalog No. 50-193-1597 Supplier Electron Microscopy Sciences Supplier No. 7205506
These disposable knives are economically priced and completely disposable. They have double honed, flexible blades with non-slip handles to ensure accurate dissection. They may be stropped for a longer lasting edge. They are available in standard or premium grades of Sheffield stainless steel.