Learn More
Electron Microscopy Sciences EMS Disposable Macro Knives Trimming Premium, 6" Blade, 0.75" width, 10.5" OAL
SDP
Catalog No. 501931599
Encompass_Preferred
These disposable knives are economically priced and completely disposable. They have double honed, flexible blades with non-slip handles to ensure accurate dissection. They may be stropped for a longer lasting edge. They are available in standard or premium grades of Sheffield stainless steel.
Blade: 6"
Width: 0.75"
OAL: 10.5"
Catalog No. 50-193-1599 Supplier Electron Microscopy Sciences Supplier No. 7205508
These disposable knives are economically priced and completely disposable. They have double honed, flexible blades with non-slip handles to ensure accurate dissection. They may be stropped for a longer lasting edge. They are available in standard or premium grades of Sheffield stainless steel.
Blade: 6"
Width: 0.75"
OAL: 10.5"