Learn More
Electron Microscopy Sciences EMS Disposable Macro Knives Brain Standard, 12" Blade, 0.75" width, 16.5" OAL
SDP
Catalog No. 501931596
Encompass_Preferred
These disposable knives are economically priced and completely disposable. They have double honed, flexible blades with non-slip handles to ensure accurate dissection. They may be stropped for a longer lasting edge. They are available in standard or premium grades of Sheffield stainless steel.
Blade: 12"
Width: 0.75"
OAL: 16.5"
Catalog No. 50-193-1596 Supplier Electron Microscopy Sciences Supplier No. 7205505
These disposable knives are economically priced and completely disposable. They have double honed, flexible blades with non-slip handles to ensure accurate dissection. They may be stropped for a longer lasting edge. They are available in standard or premium grades of Sheffield stainless steel.
Blade: 12"
Width: 0.75"
OAL: 16.5"