Learn More
Electron Microscopy Sciences Disposable Scalpel Handle Set #10
SDP
Catalog No. 501900388
Encompass_Preferred
High quality carbon steel is mounted on plastic handle, individually foil wrapped and sterilized by gamma irradiation. They are exceptionally strong and relatively uniformity sharp, ideal for routine laboratory dissecting, tissue mincing and where you do not need high cost blades.
Catalog No. 50-190-0388 Supplier Electron Microscopy Sciences Supplier No. 7205210
High quality carbon steel is mounted on plastic handle, individually foil wrapped and sterilized by gamma irradiation. They are exceptionally strong and relatively uniformity sharp, ideal for routine laboratory dissecting, tissue mincing and where you do not need high cost blades.