Learn More
Medchemexpress LLC DSR-141562 | 2007975-22-4 | 100.0% | 414.42 | 50 MG
SDP
Catalog No. 5000352260
Encompass_Preferred
DSR-141562 is a novel, orally active, and selective brain-penetrant phosphodiesterase 1 (PDE1) inhibitor. It shows preferential selectivity for human PDE1B with an IC50 of 43.9 nM, and IC50 values for human PDE1A and 1C are 97.6 and 431.8 nM, respectively. It can be used for the study of positive symptoms, negative symptoms, and cognitive impairments associated with schizophrenia.
  • Novel, orally active, and selective brain-penetrant phosphodiesterase 1 (PDE1) inhibitor.
  • Shows preferential selectivity for human PDE1B.
  • Applicable for studying positive symptoms, negative symptoms, and cognitive impairments related to schizophrenia.
Catalog No. 50-003-52260 Supplier Medchemexpress LLC Supplier No. HY13656950MG
DSR-141562 is a novel, orally active, and selective brain-penetrant phosphodiesterase 1 (PDE1) inhibitor. It shows preferential selectivity for human PDE1B with an IC50 of 43.9 nM, and IC50 values for human PDE1A and 1C are 97.6 and 431.8 nM, respectively. It can be used for the study of positive symptoms, negative symptoms, and cognitive impairments associated with schizophrenia.
  • Novel, orally active, and selective brain-penetrant phosphodiesterase 1 (PDE1) inhibitor.
  • Shows preferential selectivity for human PDE1B.
  • Applicable for studying positive symptoms, negative symptoms, and cognitive impairments related to schizophrenia.