Learn More
Medchemexpress LLC HY-136569 5mg Medchemexpress, DSR-141562 CAS:2007975-22-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1365695MG
Encompass_Preferred
Medchemexpress, HY-136569 5mg DSR-141562 CAS:2007975-22-4 DSR-141562 is a novel, orally active, and selective brain-penetrant phosphodiesterase 1 (PDE1) inhibitor. DSR-141562 shows preferential selectivity for human PDE1B with an IC50 of 43.9 nM, and the IC50 values for human PDE1A and 1C are 97.6 and 431.8 nM, respectively. DSR-141562 can be used for the study of positive symptoms, negative symptoms and cognitive impairments associated with schizophrenia[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0457