Learn More
Medchemexpress LLC DSR-6434 | 1059070-10-8 | 99.3% | 400.48 | 100 MG
SDP
Catalog No. 5000344118
Encompass_Preferred
DSR-6434 is a potent and selective Toll-like receptor 7 (TLR7) agonist, with EC50s of 7.2 nM and 4.6 nM for human and mice TLR7, respectively. It has a strong antitumor effect.
  • Potent and selective Toll-like receptor 7 agonist.
  • EC50s of 7.2 nM and 4.6 nM for human and mice TLR7.
  • Strong antitumor effect.
  • Suppresses lung metastasis in mice.
Catalog No. 50-003-44118 Supplier Medchemexpress LLC Supplier No. HY11012050MG
DSR-6434 is a potent and selective Toll-like receptor 7 (TLR7) agonist, with EC50s of 7.2 nM and 4.6 nM for human and mice TLR7, respectively. It has a strong antitumor effect.
  • Potent and selective Toll-like receptor 7 agonist.
  • EC50s of 7.2 nM and 4.6 nM for human and mice TLR7.
  • Strong antitumor effect.
  • Suppresses lung metastasis in mice.