Learn More
TARGETMOL CHEMICALS INC DSR-6434 10MG
SDP
Catalog No. 502581571
Encompass_Preferred
Also available in 5 mg 25 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DSR-6434 is a selective agonist of TLR7 with antitumor effect. DSR-6434 exhibits EC50s of 7.2 nM and 4.6 nM for human and mouse. purity: 99%
Catalog No. 50-258-1571 Supplier TARGETMOL CHEMICALS INC Supplier No. T1110810MG
Also available in 5 mg 25 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DSR-6434 is a selective agonist of TLR7 with antitumor effect. DSR-6434 exhibits EC50s of 7.2 nM and 4.6 nM for human and mouse. purity: 99%