Learn More
Cell Signaling Technology dsRNA (J2) Mouse Monoclonal Antibody, IgG2a Kappa, Unconjugated, 200 μg
SDP
Catalog No. 502731398
Encompass
The dsRNA (J2) Mouse Monoclonal Antibody is designed to detect double-stranded RNA (dsRNA) with a helix length of 40 base pairs or greater. This monoclonal antibody, produced by immunizing animals with a nucleic acid sequence, provides sequence-independent detection of dsRNA, which serves as a key marker for viral replication. It exhibits higher affinity for natural dsRNA compared to other dsRNA antigens.
  • Recognizes dsRNA helices ≥ 40 bp regardless of nucleotide composition or sequence.
  • Validated for use in immunofluorescence (immunocytochemistry) applications with a recommended dilution of 1:250 to 1:1,000.
  • Supplied as a lyophilized formulation for stable storage at -20°C.
  • Requires reconstitution in 200 μL of sterile distilled water to achieve a 1 mg/mL concentration.
  • Demonstrates reactivity across all species.
Catalog No. 50-273-1398 Supplier Cell Signaling Technology Supplier No. 76651L
The dsRNA (J2) Mouse Monoclonal Antibody is designed to detect double-stranded RNA (dsRNA) with a helix length of 40 base pairs or greater. This monoclonal antibody, produced by immunizing animals with a nucleic acid sequence, provides sequence-independent detection of dsRNA, which serves as a key marker for viral replication. It exhibits higher affinity for natural dsRNA compared to other dsRNA antigens.
  • Recognizes dsRNA helices ≥ 40 bp regardless of nucleotide composition or sequence.
  • Validated for use in immunofluorescence (immunocytochemistry) applications with a recommended dilution of 1:250 to 1:1,000.
  • Supplied as a lyophilized formulation for stable storage at -20°C.
  • Requires reconstitution in 200 μL of sterile distilled water to achieve a 1 mg/mL concentration.
  • Demonstrates reactivity across all species.