Learn More
Medchemexpress LLC HY-W019543 100mg Medchemexpress, DSS Crosslinker CAS:68528-80-3 Purity:>98%
SDP
Catalog No. 502083434
Encompass_Preferred
Medchemexpress, HY-W019543 100mg DSS Crosslinker CAS:68528-80-3 DSS Crosslinker is a non-cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3434 Supplier Medchemexpress LLC Supplier No. HYW019543100MG
Add to Cart
Add to Cart
Medchemexpress, HY-W019543 100mg DSS Crosslinker CAS:68528-80-3 DSS Crosslinker is a non-cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.