Learn More
TARGETMOL CHEMICALS INC DSS30 5MG
SDP
Catalog No. 502579519
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. DSS30 is a P25/CDK5 inhibitor.DSS30 acts by inhibiting the phosphorylation of amyloid precursor protein cleaving enzyme 1 (BACEl) which reduces the secretion of (beta)-amyloid (A(beta)).DSS30 can be used for the prevention and treatment of neurodegenerative diseases such as Alzheimer's disease. purity: 99%
Catalog No. 50-257-9519 Supplier TARGETMOL CHEMICALS INC Supplier No. T614395MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. DSS30 is a P25/CDK5 inhibitor.DSS30 acts by inhibiting the phosphorylation of amyloid precursor protein cleaving enzyme 1 (BACEl) which reduces the secretion of (beta)-amyloid (A(beta)).DSS30 can be used for the prevention and treatment of neurodegenerative diseases such as Alzheimer's disease. purity: 99%