Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dsso Crosslinker

Catalog No. 909602100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-960-2100MG 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 90-960-2100MG Supplier Sigma Aldrich Supplier No. 909602100MG
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

DSSO (disuccinimidyl sulfoxide) crosslinker is a homobifunctional, amine-targeting, sulfoxide-containing crosslinker for analysis of protein-protein interactions (PPIs) through crosslinking mass spectrometry (XL-MS). Membrane-permeable DSSO possesses two N-hydroxysuccinimide (NHS) esters for targeting Lys, a 10.1 +a spacer arm, and two symmetrical C-S cleavable bonds adjacent to the central sulfoxide. The post-cleavage spacer yields tagged peptides for unambiguous identification by collision-induced dissociation in tandem MS. DSSO Crosslinker provides complementary data to thiol-reactive and acidic residue-targeting reagents and will find wide utility in the elucidation of PPIs, study of proteins that function as complexes, quantification of structural dynamics, and the quest for targeting undruggable protein targets.Technology Spotlight: Cross-Linkers for Elucidation of Protein-Protein InteractionsDevelopment of a novel cross-linking strategy for fast and accurate identification of cross-linked peptides of protein complexesStructural dynamics of the human COP9 signalosome revealed by cross-linking mass spectrometry and integrative modelingDeveloping a Multiplexed Quantitative Cross-Linking Mass Spectrometry Platform for Comparative Structural Analysis of Protein ComplexesDevelopment of a Novel Sulfoxide-Containing MS-Cleavable Homobifunctional Cysteine-Reactive Cross-Linker for Studying Protein-Protein InteractionsDeveloping an Acidic Residue Reactive and Sulfoxide-Containing MS-Cleavable Homobifunctional Cross-Linker for Probing Protein-Protein InteractionsSubject to US Patent #9,222,943 and US Patent Application #15/275,001 of the Regents of the University of California

Specifications

CAS 361189-64-2
Density 1.068 g/mL
Molecular Formula C19H38O9
Linear Formula C19H38O9
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.