Learn More
Savillex Corporation DST-1000 Acid Purification System, Sub-Boiling Distillation, High Purity Acids, 1 L
SDP
Catalog No. NC1995627
Encompass
The DST-1000 Acid Purification System offers an efficient solution for the sub-boiling distillation of acids used in trace metal analysis. It purifies 1 ppb grade acids to ultra-pure 10 ppt grade, enhancing analytical accuracy while reducing costs compared to buying ultra-pure acids. Its air-cooled design eliminates water cooling, and it features high purity PFA construction to minimize contamination, making it ideal for sensitive laboratory applications.
Catalog No. NC1995627 Supplier Savillex Corporation Supplier No. 5251000115
Add to Cart
Add to Cart
Request a Quote
The DST-1000 Acid Purification System offers an efficient solution for the sub-boiling distillation of acids used in trace metal analysis. It purifies 1 ppb grade acids to ultra-pure 10 ppt grade, enhancing analytical accuracy while reducing costs compared to buying ultra-pure acids. Its air-cooled design eliminates water cooling, and it features high purity PFA construction to minimize contamination, making it ideal for sensitive laboratory applications.