Learn More
Medchemexpress LLC DSTYSLSSTLTLSK TFA | 80.4% | 1616.66 | 5 MG
SDP
Catalog No. 5000398593
Encompass_Preferred
DSTYSLSSTLTLSK TFA is a human peptide designed for the quantitative detection of infliximab. It is provided as a TFA salt and exhibits a peptide content of 80.4%. This product is intended for research applications only.
  • Generic human peptide
  • For infliximab quantitative detection
  • Provided as a TFA salt
  • Peptide content of 80.4%
  • For research use only
Catalog No. 50-003-98593 Supplier Medchemexpress LLC Supplier No. HYP3203A5MG
DSTYSLSSTLTLSK TFA is a human peptide designed for the quantitative detection of infliximab. It is provided as a TFA salt and exhibits a peptide content of 80.4%. This product is intended for research applications only.
  • Generic human peptide
  • For infliximab quantitative detection
  • Provided as a TFA salt
  • Peptide content of 80.4%
  • For research use only