Learn More
Medchemexpress LLC HY-112929 50mg Medchemexpress, DT-061 CAS:1809427-19-7 Purity:>98%
SDP
Catalog No. 502028829
Encompass_Preferred
Medchemexpress, HY-112929 50mg DT-061 CAS:1809427-19-7 DT-061 is an orally bioavailable activator of protein phosphatase 2A (PP2A) and could be applied in the therapy of KRAS-mutant and MYC-driven tumorigenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8829 Supplier Medchemexpress LLC Supplier No. HY11292950MG
Medchemexpress, HY-112929 50mg DT-061 CAS:1809427-19-7 DT-061 is an orally bioavailable activator of protein phosphatase 2A (PP2A) and could be applied in the therapy of KRAS-mutant and MYC-driven tumorigenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.