Learn More
Selleck Chemical LLC DT-061 S8774-10MM/1ML
SDP
Catalog No. 5000024236
Encompass_Preferred
DT-061 (SMAP) is a bioavailable PP2A activator It decreases cell viability in HCC827 and HCC3255 cell lines with IC50 values of 14 3 M and 12 4 M respectively
Catalog No. 50-000-24236 Supplier Selleck Chemical LLC Supplier No. S877410MM/1ML
DT-061 (SMAP) is a bioavailable PP2A activator It decreases cell viability in HCC827 and HCC3255 cell lines with IC50 values of 14 3 M and 12 4 M respectively