Learn More
Medchemexpress LLC DT-2 acetate | 98.6% | 3761.25 | 1 MG
SDP
Catalog No. 5000397451
Encompass_Preferred
DT-2 is a potent and selective inhibitor of the cGMP-dependent protein kinase (PKG), which inhibits PKG catalyzed phosphorylation with a Ki of 12.5 nM.
  • Potent and selective inhibitor of cGMP-dependent protein kinase (PKG).
  • Inhibits PKG catalyzed phosphorylation.
  • For research use only.
Catalog No. 50-003-97451 Supplier Medchemexpress LLC Supplier No. HYP2692B1MG
DT-2 is a potent and selective inhibitor of the cGMP-dependent protein kinase (PKG), which inhibits PKG catalyzed phosphorylation with a Ki of 12.5 nM.
  • Potent and selective inhibitor of cGMP-dependent protein kinase (PKG).
  • Inhibits PKG catalyzed phosphorylation.
  • For research use only.