Learn More
AdipoGen Bis(4-tert-Butylphenyl) iodoniumhexafluorophosphate (10 g)
SDP
Supplier:  AdipoGen CDXT0106G010
Encompass_Preferred
Chemical. CAS: 61358-25-6. Formula: C20H26F6IP. MW: 538.29. Bis(4-tert-Butylphenyl) iodoniumhexafluorophosphate is a photo-cationic polymerization initiator that is used for chemical synthesis in materials science and micro/nano electronics. Photopolymerization initiators are used in many fields to generate photocurable composites. These composites are polymerized by irradiation with UV light and electron beam which leads to altered physical properties of the composites such as solubility, viscosity and adhesiveness.
Catalog No. 50-205-4658