Learn More
Alkali Scientific DTE [Dithioerythreitol], 5 G
SDP
Catalog No. 5000661062
Encompass_Preferred
Dithioerythritol (DTE) is a reducing agent in powder form, commonly used in protein analysis and other laboratory research applications. This 5g product is ideal for reducing disulfide bonds in proteins and peptides, facilitating the analysis of their tertiary structures. DTE is often used in gel electrophoresis and chromatography to maintain proteins in their reduced state, allowing for more accurate analysis of protein interactions, folding, and structure. It is also used in biochemical assays that require the reduction of oxidative markers and can help investigate the effects of reducing agents in cellular environments.
Catalog No. 50-006-61062 Supplier Alkali Scientific Supplier No. DTE002CP
Dithioerythritol (DTE) is a reducing agent in powder form, commonly used in protein analysis and other laboratory research applications. This 5g product is ideal for reducing disulfide bonds in proteins and peptides, facilitating the analysis of their tertiary structures. DTE is often used in gel electrophoresis and chromatography to maintain proteins in their reduced state, allowing for more accurate analysis of protein interactions, folding, and structure. It is also used in biochemical assays that require the reduction of oxidative markers and can help investigate the effects of reducing agents in cellular environments.