Learn More
Medchemexpress LLC HY-138280 10mg , DTHIB CAS:897326-30-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13828010MG
Encompass_Preferred
HY-138280 10mg DTHIB CAS: 897326-30-6 DTHIB is a direct and selective heat shock factor 1 (HSF1) inhibitor with a Kd of 160 nM for DTHIB binding to the HSF1 DNA binding domain (DBD). DTHIB inhibits HSF1 cancer gene signature (HSF1 CaSig) and selectively stimulates degradation of nuclear HSF1. DTHIB has potently anticancer activities and can be used for prostate cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0810