Learn More
Selleck Chemical LLC DTHIB-5mg
SDP
Supplier:  Selleck Chemical LLC S98065MG
Encompass_Preferred
DTHIB (Direct Targeted HSF1 InhiBitor) is a direct and selective inhibitor of heat shock factor 1 (HSF1) with Kd of 160 nM for DTHIB binding to the HSF1 DNA binding domain (DBD). DTHIB exhibits potently anticancer activities.
Catalog No. 50-238-9767