Learn More
ABclonal Technology DTL Rabbit pAb
SDP
Catalog No. 5000098634
Encompass_Preferred
Contributes to ubiquitin-protein transferase activity. Involved in several processes, including protein ubiquitination, regulation of G2/M transition of mitotic cell cycle, and translesion synthesis. Located in centrosome, cytosol, and nuclear lumen. Part of Cul4A-RING E3 ubiquitin ligase complex and Cul4B-RING E3 ubiquitin ligase complex.
Catalog No. 50-000-98634 Supplier ABclonal Technology Supplier No. A1215020UL
Contributes to ubiquitin-protein transferase activity. Involved in several processes, including protein ubiquitination, regulation of G2/M transition of mitotic cell cycle, and translesion synthesis. Located in centrosome, cytosol, and nuclear lumen. Part of Cul4A-RING E3 ubiquitin ligase complex and Cul4B-RING E3 ubiquitin ligase complex.