Learn More
MSC Rust-Oleum Direct-to-Metal Acrylic Enamel 3800, Safety Yellow, Gloss, 1 gal
Catalog No. 501920050
Encompass
ROCAcrylic 3800 Direct-to-Metal Acrylic Enamel is a water-based coating suitable for both indoor and outdoor applications. This direct-to-metal enamel provides a gloss finish in safety yellow and is formulated with a low VOC content, offering excellent adhesion and protection for metal surfaces.
  • Fast drying, water based formula
  • Excellent corrosion protection
  • Ultra-smooth finished appearance
  • UV resistant for color and gloss retention
  • Dries to the touch in as little as 15 minutes
  • Can be applied in temperatures as low as 40°F
  • Provides practical coverage of 150–270 sq. ft./gal.
Catalog No. 50-192-0050 Supplier MSC Supplier No. 39035332
ROCAcrylic 3800 Direct-to-Metal Acrylic Enamel is a water-based coating suitable for both indoor and outdoor applications. This direct-to-metal enamel provides a gloss finish in safety yellow and is formulated with a low VOC content, offering excellent adhesion and protection for metal surfaces.
  • Fast drying, water based formula
  • Excellent corrosion protection
  • Ultra-smooth finished appearance
  • UV resistant for color and gloss retention
  • Dries to the touch in as little as 15 minutes
  • Can be applied in temperatures as low as 40°F
  • Provides practical coverage of 150–270 sq. ft./gal.