Learn More
Medchemexpress LLC DTME 5mg
SDP
Catalog No. 5000205463
Encompass_Preferred
DTME (dithio-bis-maleimidoethane) is a homobifunctional maleimide crosslinker specifically designed for conjugation between sulfhydryl groups (-SH) DTME whose molecular structure consists of two maleimide groups connected by an ethylene disulfide bridge can specifically react with thiol - containing molecules (such as cysteine residues) to form stable covalent bonds DTME allows crosslinks that can be cleaved with reducing agents such as DTT (HY-15917) DTME is commonly utilized to explore and characterize protein structure particularly oligomerization or protein interactions[1]
Catalog No. 50-002-05463 Supplier Medchemexpress LLC Supplier No. HYW1064115MG
DTME (dithio-bis-maleimidoethane) is a homobifunctional maleimide crosslinker specifically designed for conjugation between sulfhydryl groups (-SH) DTME whose molecular structure consists of two maleimide groups connected by an ethylene disulfide bridge can specifically react with thiol - containing molecules (such as cysteine residues) to form stable covalent bonds DTME allows crosslinks that can be cleaved with reducing agents such as DTT (HY-15917) DTME is commonly utilized to explore and characterize protein structure particularly oligomerization or protein interactions[1]