Learn More
AdipoGen DTNB, 69-78-3, MFCD00007140, 1 g
SDP
Catalog No. 501147015
Encompass_Preferred
MF: C14H8N2O8S2, MW: 396.35, ≥98%, Appearance: Yellow powder. A sensitive reagent for measuring the free sulfhydryl content in proteins, peptides, and tissues. Used to characterize reactive thiol groups and photometric determination of thiols and for measuring low-molecular mass thiols such as glutathione in both pure solutions and biological samples, such as blood. It can also measure the number of thiol groups on proteins.
Catalog No. 50-114-7015 Supplier AdipoGen Supplier No. CDXD0156G001
MF: C14H8N2O8S2, MW: 396.35, ≥98%, Appearance: Yellow powder. A sensitive reagent for measuring the free sulfhydryl content in proteins, peptides, and tissues. Used to characterize reactive thiol groups and photometric determination of thiols and for measuring low-molecular mass thiols such as glutathione in both pure solutions and biological samples, such as blood. It can also measure the number of thiol groups on proteins.