Learn More
Selleck Chemical LLC DTNB S2764-500mg
SDP
Catalog No. 503108158
Encompass_Preferred
DTNB (Ellman s Reag Ellmans Reagenz 5 5 -Dithiobis(2-nitrobenzoic acid) 5 5 -Dithiobis-2-nitrobenzoes ure) is a non-fluorescent probe used to quantify the number or concentration of thiol groups in a sample DTNB is also an allosteric inhibitor of dengue virus protease (NS2B-NS3pro) and Streptomyces proteases
Catalog No. 50-310-8158 Supplier Selleck Chemical LLC Supplier No. S2764500MG
DTNB (Ellman s Reag Ellmans Reagenz 5 5 -Dithiobis(2-nitrobenzoic acid) 5 5 -Dithiobis-2-nitrobenzoes ure) is a non-fluorescent probe used to quantify the number or concentration of thiol groups in a sample DTNB is also an allosteric inhibitor of dengue virus protease (NS2B-NS3pro) and Streptomyces proteases