Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DTNB; Ellman's Reagent, 5,5'-Dithiobis-(2-Nitrobenzoic Acid)

Catalog No. D8451
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D8451 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D8451 Supplier Invitrogen™ Supplier No. D8451
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

DTNB; Ellman's Reagent, 5,5'-Dithiobis-(2-Nitrobenzoic Acid)

DTNB or Elman's reagent can be used to quantitate thiols in proteins, cells and plasma by absorption measurements. It readily forms a mixed disulfide with thiols, liberating the chromophore 5-merapto-2-nitrobenzoic acid (absorption maximum 410 nm). Only protein thiols that are accessible to this water-soluble reagent are modified.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Elman's Reagent
Quantity 10 g
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.