Learn More
Perkin Elmer US LLC D-Torch 1-Slot Quartz Outer Tube for Avio 200/500
Catalog No. 502117274
Encompass_Preferred
This Outer Tube is included in the D-Torch Complete Assembly with Quartz Outer Tube for Avio 200 (N0790442) and the D-Torch Complete Assembly with Quartz Outer Tube for Avio 500 (N0810861). Features and Benefits of the D-Torch include: Analysis of high-matrix solutions (i.e. fusion samples, samples with high-dissolved solids, samples with high-acid content) which can cause early devitrification or sample deposition when using quartz Demountable outer tube With the D-Torch, you need only replace the outer tube Interchangeable quartz and ceramic outer tubes (N0791303, N0811912) Interchangeable injectors choose the optimum injector for your application. Select from quartz for aqueous solutions, narrow bore for organics, wide bore for high dissolved solids or ceramic for HF
Catalog No. 50-211-7274 Supplier Perkin Elmer US LLC Supplier No. N0791301
This Outer Tube is included in the D-Torch Complete Assembly with Quartz Outer Tube for Avio 200 (N0790442) and the D-Torch Complete Assembly with Quartz Outer Tube for Avio 500 (N0810861). Features and Benefits of the D-Torch include: Analysis of high-matrix solutions (i.e. fusion samples, samples with high-dissolved solids, samples with high-acid content) which can cause early devitrification or sample deposition when using quartz Demountable outer tube With the D-Torch, you need only replace the outer tube Interchangeable quartz and ceramic outer tubes (N0791303, N0811912) Interchangeable injectors choose the optimum injector for your application. Select from quartz for aqueous solutions, narrow bore for organics, wide bore for high dissolved solids or ceramic for HF