Learn More
Perkin Elmer US LLC D-Torch Base and Inner Tube for Optima 2x00/4x00/5x00/7x00 DV
Catalog No. 502105258
Encompass_Preferred
The D-Torch provides a demountable torch design without sacrificing performance or usability. Featuring a torch body with fixed alumina intermediate tube for greater robustness, in addition to the option for interchangable outer tubes and injectors, the D-Torch significantly reduces torch replacement costs for the analysis of challenging sample matrices. It also features an O-ring free torch body for simplified maintenance and is compatible with all commonly used spray chambers. Only the Quartz Outer Tube (N0777510) is compatible with this Base and Inner Tube assembly for Optima 2x00/4x00/5x00/7x00 DV.
Catalog No. 50-210-5258 Supplier Perkin Elmer US LLC Supplier No. N0777509
The D-Torch provides a demountable torch design without sacrificing performance or usability. Featuring a torch body with fixed alumina intermediate tube for greater robustness, in addition to the option for interchangable outer tubes and injectors, the D-Torch significantly reduces torch replacement costs for the analysis of challenging sample matrices. It also features an O-ring free torch body for simplified maintenance and is compatible with all commonly used spray chambers. Only the Quartz Outer Tube (N0777510) is compatible with this Base and Inner Tube assembly for Optima 2x00/4x00/5x00/7x00 DV.