Learn More
Perkin Elmer US LLC D-Torch Body for Avio 500
Catalog No. 502113151
Encompass_Preferred
The D-Torch Body is included in the D-Torch Complete Assembly with Quartz Outer Tube (N0810861). Features and Benefits of the D-Torch include: Analysis of high-matrix solutions (i.e. fusion samples, samples with high-dissolved solids, samples with high-acid content) which can cause early devitrification or sample deposition when using quartz Demountable outer tube With the D-Torch, you need only replace the outer tube Interchangeable quartz and ceramic outer tubes (N0811912) Interchangeable injectors choose the optimum injector for your application. Select from quartz for aqueous solutions, narrow bore for organics, wide bore for high dissolved solids or ceramic for HF
Catalog No. 50-211-3151 Supplier Perkin Elmer US LLC Supplier No. N0811904
The D-Torch Body is included in the D-Torch Complete Assembly with Quartz Outer Tube (N0810861). Features and Benefits of the D-Torch include: Analysis of high-matrix solutions (i.e. fusion samples, samples with high-dissolved solids, samples with high-acid content) which can cause early devitrification or sample deposition when using quartz Demountable outer tube With the D-Torch, you need only replace the outer tube Interchangeable quartz and ceramic outer tubes (N0811912) Interchangeable injectors choose the optimum injector for your application. Select from quartz for aqueous solutions, narrow bore for organics, wide bore for high dissolved solids or ceramic for HF