Learn More
Medchemexpress LLC HY-100538A 10mg Medchemexpress, DTP3 (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100538A10MG
Encompass_Preferred
Medchemexpress, HY-100538A 10mg DTP3 (TFA) CAS: DTP3 TFA is a potent and selective GADD45β/MKK7 (growth arrest and DNA-damage-inducible β/mitogen-activated protein kinase kinase 7) inhibitor. DTP3 TFA targets an essential, cancer-selective cell-survival module downstream of the NF-κB pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7742