Learn More
Apexbio Technology LLC DTP3 TFA 1809784-29-9 (free-base) 25mg
SDP
Catalog No. 5000090433
Encompass_Preferred
DTP3 TFA is a small-molecule inhibitor targeting the GADD45 /MKK7 protein-protein interaction complex It is designed to disrupt this interaction thereby modulating downstream NF- B signaling involved in tumor cell survival DTP3 TFA exerts its biological activity primarily by binding directly to MKK7 (KD 64 81 nM) inducing conformational changes that dissociate GADD45 from MKK7 resulting in activation of JNK signaling In cell-based studies DTP3 TFA selectively inhibits proliferation of cancer cell lines with elevated GADD45 expression Based on these pharmacological properties DTP3 TFA holds research potential in the study and therapeutic intervention of cancers including multiple myeloma via disruption of GADD45 /MKK7 signaling
Catalog No. 50-000-90433 Supplier Apexbio Technology LLC Supplier No. B488625
DTP3 TFA is a small-molecule inhibitor targeting the GADD45 /MKK7 protein-protein interaction complex It is designed to disrupt this interaction thereby modulating downstream NF- B signaling involved in tumor cell survival DTP3 TFA exerts its biological activity primarily by binding directly to MKK7 (KD 64 81 nM) inducing conformational changes that dissociate GADD45 from MKK7 resulting in activation of JNK signaling In cell-based studies DTP3 TFA selectively inhibits proliferation of cancer cell lines with elevated GADD45 expression Based on these pharmacological properties DTP3 TFA holds research potential in the study and therapeutic intervention of cancers including multiple myeloma via disruption of GADD45 /MKK7 signaling