Learn More
Medchemexpress LLC DTPA anhydride | 23911-26-4 | ≥97.0% | 357.32 | 5 G
SDP
Catalog No. 5000452080
Encompass_Preferred
DTPA anhydride is a bifunctional chelator. Its anhydride can react with amino groups in proteins (such as lysine residues) to form stable amide bonds. It can also bind to radionuclides to synthesize radionuclide-labeled drug conjugates (RDCs), which have the ability to specifically target biomolecules and can be used in medical imaging or therapy.
  • Bifunctional chelator
  • Anhydride reacts with amino groups in proteins to form stable amide bonds
  • Binds to radionuclides to synthesize radionuclide-labeled drug conjugates (RDCs)
  • RDCs specifically target biomolecules
  • Can be used in medical imaging or therapy
Catalog No. 50-004-52080 Supplier Medchemexpress LLC Supplier No. HYW0111185G
DTPA anhydride is a bifunctional chelator. Its anhydride can react with amino groups in proteins (such as lysine residues) to form stable amide bonds. It can also bind to radionuclides to synthesize radionuclide-labeled drug conjugates (RDCs), which have the ability to specifically target biomolecules and can be used in medical imaging or therapy.
  • Bifunctional chelator
  • Anhydride reacts with amino groups in proteins to form stable amide bonds
  • Binds to radionuclides to synthesize radionuclide-labeled drug conjugates (RDCs)
  • RDCs specifically target biomolecules
  • Can be used in medical imaging or therapy