Learn More
Medchemexpress LLC DTPA anhydride | 23911-26-4 | ≥97.0% | 357.32 | 50 G
SDP
Catalog No. 5000453576
Encompass_Preferred
DTPA anhydride is a bifunctional chelator. Its anhydride can react with amino groups in proteins (such as lysine residues) to form stable amide bonds. It can also bind to radionuclides to synthesize radionuclide-labeled drug conjugates (RDCs). RDCs have the ability to specifically target biomolecules and can be used in medical imaging or therapy.
  • Bifunctional chelator
  • Reacts with amino groups in proteins to form stable amide bonds
  • Binds to radionuclides to synthesize radionuclide-labeled drug conjugates
  • Drug conjugates can specifically target biomolecules for medical imaging or therapy
Catalog No. 50-004-53576 Supplier Medchemexpress LLC Supplier No. HYW01111850G
Add to Cart
Add to Cart
DTPA anhydride is a bifunctional chelator. Its anhydride can react with amino groups in proteins (such as lysine residues) to form stable amide bonds. It can also bind to radionuclides to synthesize radionuclide-labeled drug conjugates (RDCs). RDCs have the ability to specifically target biomolecules and can be used in medical imaging or therapy.
  • Bifunctional chelator
  • Reacts with amino groups in proteins to form stable amide bonds
  • Binds to radionuclides to synthesize radionuclide-labeled drug conjugates
  • Drug conjugates can specifically target biomolecules for medical imaging or therapy