Learn More
Medchemexpress LLC DTPA-tetra (t-Bu ester) | 174267-71-1 | 99.0% | 617.77 g·mol⁻¹ | C30H55N3O10 | 10 MG
SDP
Catalog No. 5000297911
Encompass_Preferred
DTPA-tetra (t-Bu ester) is the tetra-tert-butyl ester derivative of diethylenetriaminepentaacetic acid (DTPA). It serves as a bifunctional chelator for use in bioconjugation and radiolabeling workflows, enabling conjugation of peptides and radionuclides for labeling and tumor pre-targeting research applications.
  • Bifunctional chelator for peptide and radionuclide conjugation.
  • Tetra-tert-butyl ester derivative that facilitates protection and conjugation chemistry.
  • High purity (≈99.0%) suitable for research applications.
  • Molecular weight 617.77 g·mol⁻¹ and molecular formula C30H55N3O10.
  • Supplied as small quantities for labeling and method development workflows.
Catalog No. 50-002-97911 Supplier Medchemexpress LLC Supplier No. HYA007410MG
DTPA-tetra (t-Bu ester) is the tetra-tert-butyl ester derivative of diethylenetriaminepentaacetic acid (DTPA). It serves as a bifunctional chelator for use in bioconjugation and radiolabeling workflows, enabling conjugation of peptides and radionuclides for labeling and tumor pre-targeting research applications.
  • Bifunctional chelator for peptide and radionuclide conjugation.
  • Tetra-tert-butyl ester derivative that facilitates protection and conjugation chemistry.
  • High purity (≈99.0%) suitable for research applications.
  • Molecular weight 617.77 g·mol⁻¹ and molecular formula C30H55N3O10.
  • Supplied as small quantities for labeling and method development workflows.