Learn More
Medchemexpress LLC Dtpa-tetra (t-bu ester) | 174267-71-1 | 99.0% | C30H55N3O10 | 5 MG
SDP
Catalog No. 5000432126
Encompass_Preferred
DTPA-tetra (t-Bu ester) is a bifunctional chelator (BFC) and a macrocyclic DPTA derivative. It is used for tumor pre-targeting and can be utilized for the conjugation of peptides and radionuclides. It is for research use only.
  • Bifunctional chelator (BFC)
  • Macrocyclic DPTA derivative
  • Used for tumor pre-targeting
  • Can be utilized for the conjugation of peptides and radionuclides
Catalog No. 50-004-32126 Supplier Medchemexpress LLC Supplier No. HYA00745MG
Add to Cart
Add to Cart
DTPA-tetra (t-Bu ester) is a bifunctional chelator (BFC) and a macrocyclic DPTA derivative. It is used for tumor pre-targeting and can be utilized for the conjugation of peptides and radionuclides. It is for research use only.
  • Bifunctional chelator (BFC)
  • Macrocyclic DPTA derivative
  • Used for tumor pre-targeting
  • Can be utilized for the conjugation of peptides and radionuclides