Learn More
Medchemexpress LLC DTPA-tetra (t-Bu ester) | 174267-71-1 | 99.0% | C30H55N3O10 | 50 MG
SDP
Catalog No. 5000432333
Encompass_Preferred
DTPA-tetra (t-Bu ester) is a bifunctional chelator (BFC) and a macrocyclic DPTA derivative used for tumor pre-targeting. It can be used for conjugation of peptides and radionuclides.
  • Bifunctional chelator (BFC)
  • Macrocyclic DPTA derivative
  • Used for tumor pre-targeting
  • Can be used for conjugation of peptides
  • Can be used for conjugation of radionuclides
Catalog No. 50-004-32333 Supplier Medchemexpress LLC Supplier No. HYA007450MG
DTPA-tetra (t-Bu ester) is a bifunctional chelator (BFC) and a macrocyclic DPTA derivative used for tumor pre-targeting. It can be used for conjugation of peptides and radionuclides.
  • Bifunctional chelator (BFC)
  • Macrocyclic DPTA derivative
  • Used for tumor pre-targeting
  • Can be used for conjugation of peptides
  • Can be used for conjugation of radionuclides