Learn More
Medchemexpress LLC DTPD-Q | 252950-56-4 | 99.7% | 318.37 | 25 MG
SDP
Catalog No. 5000393268
Encompass_Preferred
DTPD-Q is a transformation product of the tire additive N,N′-diphenyl-p-phenylenediamine (DTPD). It accumulates and increases ecological risk in the environment.
  • Inhibits dynamin 1 (IC50 = 273 μM).
  • Reduces clathrin-mediated endocytosis in serum-starved U2OS cells (IC50 = 120 μM).
  • Less toxic to the aquatic bacterium V. fischeri than DTPD (EC50 = 1.98 mg/L).
  • Increases intestinal permeability and the production of reactive oxygen species (ROS) in C. elegans at concentrations of 1 or 10 μg/ml.
  • Detected in roadway runoff water, roadside soils, indoor dust, and consumer products made from recycled tire rubber.
Catalog No. 50-003-93268 Supplier Medchemexpress LLC Supplier No. 25MGHY163650
DTPD-Q is a transformation product of the tire additive N,N′-diphenyl-p-phenylenediamine (DTPD). It accumulates and increases ecological risk in the environment.
  • Inhibits dynamin 1 (IC50 = 273 μM).
  • Reduces clathrin-mediated endocytosis in serum-starved U2OS cells (IC50 = 120 μM).
  • Less toxic to the aquatic bacterium V. fischeri than DTPD (EC50 = 1.98 mg/L).
  • Increases intestinal permeability and the production of reactive oxygen species (ROS) in C. elegans at concentrations of 1 or 10 μg/ml.
  • Detected in roadway runoff water, roadside soils, indoor dust, and consumer products made from recycled tire rubber.