Learn More
Medchemexpress LLC DTPD-Q | 252950-56-4 | 99.7% | 318.37 | 5 MG
SDP
Catalog No. 5000392531
Encompass_Preferred
DTPD-Q is a transformation product of the tire additive N,N′-diphenyl-p-phenylenediamine (DTPD). It accumulates and increases ecological risk in the environment.
  • For research use only.
  • Appearance: Solid.
  • Color: Yellow to brown.
Catalog No. 50-003-92531 Supplier Medchemexpress LLC Supplier No. HY1636505MG
DTPD-Q is a transformation product of the tire additive N,N′-diphenyl-p-phenylenediamine (DTPD). It accumulates and increases ecological risk in the environment.
  • For research use only.
  • Appearance: Solid.
  • Color: Yellow to brown.