Learn More
Medchemexpress LLC dTRIM24 | 2170695-14-2 | 99.89% | 1113.30 | 10 MG
SDP
Catalog No. 501968900
Encompass_Preferred
dTRIM24 is a selective bifunctional degrader of TRIM24, designed to suppress growth in MOLM-13 cells by recruiting the VHL E3 ubiquitin ligase. This PROTAC-based degrader leads to potent and selective degradation of TRIM24, causing apoptosis and a sustained proliferative defect.
  • Potent and selective degradation of TRIM24
  • Suppresses growth in MOLM-13 cells
  • Induces apoptosis with enhanced PARP cleavage
  • Based on PROTAC technology
Catalog No. 50-196-8900 Supplier Medchemexpress LLC Supplier No. HY11151910MG
dTRIM24 is a selective bifunctional degrader of TRIM24, designed to suppress growth in MOLM-13 cells by recruiting the VHL E3 ubiquitin ligase. This PROTAC-based degrader leads to potent and selective degradation of TRIM24, causing apoptosis and a sustained proliferative defect.
  • Potent and selective degradation of TRIM24
  • Suppresses growth in MOLM-13 cells
  • Induces apoptosis with enhanced PARP cleavage
  • Based on PROTAC technology