Learn More
CPC Scientific Glp-His-Trp-Ser-Tyr-DTrp-Leu-Arg-Pro-Gly-NH2 (acetate salt) 1MG
SDP
Supplier:  CPC Scientific LHRH004A
Encompass
LHRH agonist used in the treatment of prostate cancer.ONE-LETTER SEQUENCE: Glp-HWSY-DTrp-LRPG-NH2MOLECULAR FORMULA: C64H82N18O13MOLECULAR WEIGHT:1311.47STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [57773-63-4], SYNONYMS: (D-Trp)-GnRH, Triptorelin acetate salt, (D-Trp)-LHRHRESEARCH AREA: Hormonal
Catalog No. 50-210-9650