Learn More
Malvern Panalytical Folded Capillary Zeta Cell, pack of 10 with 20 stoppers
Catalog No. NC2035003
Encompass
Disposable cuvettes primarily for the measurement of zeta potential with the Zetasizer Nano & Zetasizer Advance ranges, but can be used for size measurement. Pack of 10 cells with 20 stoppers. Requires Zetasizer software version 7.02 or later for Nano Range.
Catalog No. NC2035003 Supplier Malvern Panalytical Supplier No. 10038296
Disposable cuvettes primarily for the measurement of zeta potential with the Zetasizer Nano & Zetasizer Advance ranges, but can be used for size measurement. Pack of 10 cells with 20 stoppers. Requires Zetasizer software version 7.02 or later for Nano Range.