Learn More
Supply Solutions DTT, 3483-12-3, MFCD00004877, 10g
Catalog No. 501005150
Encompass
Linear Formula HSCH2CH(OH)CH(OH)CH2SH, Molecular Weight 154.25, Synonym: 1,4-dithiothreitol, threo-1,4-Dimercapto-2,3-butanediol, DL-Dithiothreitol, Clelands reagent, DTT, cleland?s reagent, dithiothreitol; 1,4-dithiothreitol is commonly known as Cleland?s reagent. It confers protection to thiol groups and reduces disulfide bonds in peptides and proteins.[
Catalog No. 50-100-5150 Supplier Supply Solutions Supplier No. SIGMA 10708984001
Linear Formula HSCH2CH(OH)CH(OH)CH2SH, Molecular Weight 154.25, Synonym: 1,4-dithiothreitol, threo-1,4-Dimercapto-2,3-butanediol, DL-Dithiothreitol, Clelands reagent, DTT, cleland?s reagent, dithiothreitol; 1,4-dithiothreitol is commonly known as Cleland?s reagent. It confers protection to thiol groups and reduces disulfide bonds in peptides and proteins.[