Learn More
Enzo Life Sciences DTT (5g). CAS: 3483-12-3
SDP
Catalog No. 502008633
Encompass_Preferred
Cell permeable protective agent for SH groups, maintaining monothiols completely in the reduced state and of reducing disulfides quantitatively. ER-stress inducer. Alternative name: 1,4-Dithio-DL-threitol, DL-Dithiothreitol, Cleland's Reagent. Purity: ≥98% (Assay by Iodometric Titration). Solubility: Soluble in water, 100% ethanol, acetone or ethyl acetate. Long Term Storage: +4°C.
Catalog No. 50-200-8633 Supplier Enzo Life Sciences Supplier No. ALX280001G005
Cell permeable protective agent for SH groups, maintaining monothiols completely in the reduced state and of reducing disulfides quantitatively. ER-stress inducer. Alternative name: 1,4-Dithio-DL-threitol, DL-Dithiothreitol, Cleland's Reagent. Purity: ≥98% (Assay by Iodometric Titration). Solubility: Soluble in water, 100% ethanol, acetone or ethyl acetate. Long Term Storage: +4°C.