Learn More
Research Products International Corp DTT [DL-Dithiothreitol] (Cleland's Reagent)], 100 Grams
SDP
Supplier:  Research Products International Corp D11000100.0
Encompass_Preferred
Protective reagent for maintaining -SH groups in reduced state. DTT permeates the cell membrane protecting protein sulfhydryls thereby restoring enzyme activity lost by oxidation of the groups.
Catalog No. 50-213-270