Learn More
Research Products International Corp DTT [DL-Dithiothreitol] (Cleland's Reagent)], 25 Grams
SDP
Catalog No. 50213271
Encompass_Preferred
Protective reagent for maintaining -SH groups in reduced state. DTT permeates the cell membrane protecting protein sulfhydryls thereby restoring enzyme activity lost by oxidation of the groups.
Catalog No. 50-213-271 Supplier Research Products International Corp Supplier No. D1100025.0
Protective reagent for maintaining -SH groups in reduced state. DTT permeates the cell membrane protecting protein sulfhydryls thereby restoring enzyme activity lost by oxidation of the groups.