Learn More
Gold Biotechnology Inc DTT (Dithiothreitol) (> 99% pure) Protease free 50 g
SDP
Supplier:  Gold Biotechnology Inc DTT50
Encompass_Preferred
Dithiothreitol (DTT) is the common name of the more popular of the two Cleland’s Reagents (the other being Dithioerythritol or DTE). DTT is a powerful reducing agent that forms a stable six-membered ring with an internal disulfide bond which is resistant to oxidation. DTT is often used for the following: reducing the disulfide bridge of the cross-linker N,N′-bis(acryloyl) cystamine to break apart the matrix of a polyacrylamide gel, the reduction of disulfide bonds in proteins, the prevention of those bonds from forming between cysteine residues and lastly, DTT is often used to reduce thiolated DNA in order to minimize dimerization.
Catalog No. 50-153-2769