Learn More
Alkali Scientific DTT [DL Dithiothreitol] (Cleland's Reagent)], 10 G
SDP
Catalog No. 5000660945
Encompass_Preferred
Dithiothreitol (DTT), also known as Cleland's Reagent, is a highly effective reducing agent used in protein stabilization and enzyme assays. This 10g reagent is utilized in laboratory applications that require the reduction of disulfide bonds to prevent oxidation and preserve protein function. DTT is essential in maintaining the integrity of proteins during purification, electrophoresis, and structural studies. It is also used in cellular assays to protect proteins from oxidative damage and facilitate accurate enzyme activity measurements. DTT is an indispensable tool in molecular biology, biochemistry, and pharmaceutical research where maintaining protein structure and function is crucial.
Catalog No. 50-006-60945 Supplier Alkali Scientific Supplier No. DTT002CP
Dithiothreitol (DTT), also known as Cleland's Reagent, is a highly effective reducing agent used in protein stabilization and enzyme assays. This 10g reagent is utilized in laboratory applications that require the reduction of disulfide bonds to prevent oxidation and preserve protein function. DTT is essential in maintaining the integrity of proteins during purification, electrophoresis, and structural studies. It is also used in cellular assays to protect proteins from oxidative damage and facilitate accurate enzyme activity measurements. DTT is an indispensable tool in molecular biology, biochemistry, and pharmaceutical research where maintaining protein structure and function is crucial.